Methyl-beta-D-galactopyranoside 98%

供应商: Thermo Fisher Scientific

Synonyms: Methyl-β-D-galactoside, (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol, (2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol

314980050 314980010
ACRO314980050EA 0 CNY
ACRO314980050 ACRO314980010
Methyl-beta-D-galactopyranoside 98%
Methyl-beta-D-galactopyranoside

CAS No.: 1824-94-8

分子式: C₇H₁₄O₆
分子量: 194.18 g/mol
溶点: 175…179 °C
存储条件: Refrigerator
MDL编号: MFCD00064357
CAS号: 1824-94-8

现在订购

产品技术规格

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
HPLC ≥97.5 % (ELSD)
Specific optical rotation -16° to -20° (20°C, 589 nm) (c=1.5, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR