1,2-Bis(chlorodimethylsilyl)ethane 96%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2,5-二氯-2,5-二甲基-2,5-二硅杂己烷, 1,1,4,4-四甲基-1,4-二氯乙烯乙硅烷
313260050
313260250
ACRO313260050EA
0
CNY
ACRO313260050
ACRO313260250
1,2-Bis(chlorodimethylsilyl)ethane 96%
1,2-双(氯二甲基硅基)乙烷
Appearance: Transparent Low melting solid
分子式:
C₆H₁₆Cl₂Si₂ 分子量: 215.26 g/mol 沸点: 198…200 °C (760 mmHg) 溶点: 35…38 °C 闪点: 65 °C 存储条件: Ambient |
MDL编号:
MFCD00000505 CAS号: 13528-93-3 UN号: 3261 ADR: 8 II |
产品技术规格
Appearance | Transparent Low melting solid |
Infrared spectrum | Conforms |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...