(S)-5-(羟甲基)-2-吡咯烷酮 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: L-焦谷氨醇

303140010 303140050
ACRO303140010EA 0 CNY
ACRO303140010 ACRO303140050
(S)-5-(羟甲基)-2-吡咯烷酮 97%
(S)-5-(羟甲基)-2-吡咯烷酮

CAS No.: 17342-08-4

分子式: C₅H₉NO₂
分子量: 115.13 g/mol
溶点: 78…86 °C
存储条件: Ambient
MDL编号: MFCD00077792
CAS号: 17342-08-4

现在订购

产品技术规格

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 78 to 86 °C
HPLC ≥96.0 %
Specific optical rotation +27.5° to +32.5° (20 °C, 589 nm) (c=2, C2H5OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR