Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: Potassium Hydrotris(3,5-dimethylpyrazol-1-yl)borate, Potassium hydrotris(3,5-dimethylpyrazol-1-yl)borate

296020050
ACRO296020050EA 0 CNY
ACRO296020050
Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride 99%
Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride

Appearance: White Powder or needles

分子式: C₁₅H₂₂BKN₆
分子量: 336.29 g/mol
溶点: 287 °C
存储条件: Ambient
MDL编号: MFCD00040342
CAS号: 17567-17-8
UN号: 2813
ADR: 4.3 III

现在订购

产品技术规格

Appearance White Powder or needles
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR