2-Amino-6-hydroxy-8-mercaptopurine 97%
供应商: Thermo Fisher Scientific
|
Warning
|
295230050
295230250
ACRO295230050EA
0
CNY
ACRO295230050
ACRO295230250
2-Amino-6-hydroxy-8-mercaptopurine 97%
2-Amino-6-hydroxy-8-mercaptopurine
Appearance: Yellow Crystalline powder
分子式:
C₅H₅N₅OS 溶点: >300 °C 存储条件: Ambient |
MDL编号:
MFCD00075623 CAS号: 28128-40-7 |
产品技术规格
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...