2-(Pyridin-4-yl)acetic acid hydrochloride 98%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-Pyridylacetic acid hydrochloride
294750050
294750010
ACRO294750050EA
0
CNY
ACRO294750050
ACRO294750010
2-(Pyridin-4-yl)acetic acid hydrochloride 98%
2-(Pyridin-4-yl)acetic acid hydrochloride
Appearance: White to cream Crystalline powder or crystals and/or chunks
分子式:
C₇H₈ClNO₂ 分子量: 173.60 g/mol 溶点: 139…141 °C 存储条件: Ambient |
MDL编号:
MFCD00012827 CAS号: 6622-91-9 |
产品技术规格
Appearance | White to cream Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...