3-环己烯-1-甲酸 97%

供应商: Thermo Fisher Scientific
Danger

294390100 294390010
ACRO294390100EA 0 CNY
ACRO294390100 ACRO294390010
3-环己烯-1-甲酸 97%
3-环己烯-1-甲酸

Appearance: Clear yellow-brown to blue-grey Liquid

分子式: C₇H₁₀O₂
沸点: 238 °C (760 mmHg)
溶点: 17 °C
密度: 1,08 g/cm³
闪点: >112 °C
存储条件: Ambient
MDL编号: MFCD00013781
CAS号: 4771-80-6
UN号: 3265
ADR: 8,II

现在订购

产品技术规格

Appearance Clear yellow-brown to blue-grey Liquid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4820 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR