双(二亚苄基丙酮)钯(0)

供应商: Thermo Fisher Scientific
Warning

Synonyms: 双(二苄亚基丙酮)钯(0)

291970010 291970050
ACRO291970010EA 0 CNY
ACRO291970010 ACRO291970050
双(二亚苄基丙酮)钯(0)
双(二亚苄基丙酮)钯(0)

Appearance: Red-brown to black Powder

分子式: C₃₄H₂₈O₂Pd
溶点: 150 °C
存储条件: Ambient
MDL编号: MFCD00051942
CAS号: 32005-36-0

现在订购

产品技术规格

Appearance Red-brown to black Powder
Assay 16 to 21 % (Pd)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR