Boc-Sar-OH 99+%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
BOC-肌氨酸, Boc-N-甲基甘氨酸
275670050
275670250
ACRO275670050EA
0
CNY
ACRO275670050
ACRO275670250
Boc-Sar-OH 99+%
Boc-Sar-OH
Appearance: White Crystalline powder or crystals
分子式:
C₈H₁₅NO₄ 分子量: 189.211 g/mol 溶点: 86…90 °C 存储条件: Refrigerator |
MDL编号:
MFCD00037795 CAS号: 13734-36-6 |
产品技术规格
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 86 to 90 °C |
Water | ≤0.2 % (K.F.) |
HPLC | ≥99.0 % (ELSD) |
Titration with NaOH | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...