三丙酸甘油酯 97%

供应商: Thermo Fisher Scientific

Synonyms: 甘油三丙酸酯

275101000 275102500
ACRO275101000EA 0 CNY
ACRO275101000 ACRO275102500
三丙酸甘油酯 97%
三丙酸甘油酯

Appearance: Clear colorless Liquid

分子式: C₁₂H₂₀O₆
分子量: 260.29 g/mol
沸点: 151 °C (2 mmHg)
密度: 1.082 g/cm³
闪点: 162 °C
存储条件: Ambient
MDL编号: MFCD00027005
CAS号: 139-45-7

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4340 (20°C, 589 nm)
GC ≥96.0 %
Specific gravity (20/20°C) 1.082 to 1.088


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR