反-玉米素 (synthetic) 97+%
供应商: Thermo Fisher Scientific
264290100
264291000
ACRO264290100EA
0
CNY
ACRO264290100
ACRO264291000
反-玉米素 (synthetic) 97+%
反-玉米素
Appearance: White to cream Powder
分子式:
C₁₀H₁₃N₅O 溶点: 207 °C 存储条件: Refrigerator |
MDL编号:
MFCD00213654 CAS号: 1637-39-4 |
产品技术规格
Appearance | White to cream Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Melting point | ≥207°C |
Loss on drying | ≤1 % |
HPLC | ≥97.0 % |
Related substances | ≤2 % (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...