DL-Selenomethionine 99+%

供应商: Thermo Fisher Scientific
Danger

Synonyms: H-DL-Se-Met-OH, 2-Amino-4-(methylseleno)butyric Acid, 2-Amino-4-(methylselanyl)butanoic acid

240770010 240770025 240775000
ACRO240770010EA 0 CNY
ACRO240770010 ACRO240770025 ACRO240775000
DL-Selenomethionine 99+%
DL-Selenomethionine

Appearance: Colorless Glistening powder

分子式: C₅H₁₁NO₂Se
分子量: 196.11 g/mol
溶点: 265 °C
存储条件: Refrigerator
MDL编号: MFCD00063089
CAS号: 1464-42-2
UN号: 3283
ADR: 6.1 III

现在订购

产品技术规格

Appearance Colorless to white to off-white Glistening powder or flakes
Infrared spectrum Conforms
HPLC ≥99.0 %
Specific optical rotation -0.5° to +0.5° (20°C, 589 nm) (c=1, 1 N HCl)
Titration with HClO4 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR