2,6-二氨基庚二酸 95%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2,6-二氨基蒲桃酸
235540010
235540050
235540250
ACRO235540010EA
0
CNY
ACRO235540010
ACRO235540050
ACRO235540250
2,6-二氨基庚二酸 95%
2,6-二氨基庚二酸
CAS No.: 583-93-7
分子式:
C₇H₁₄N₂O₄ 分子量: 190.20 g/mol 溶点: 300 °C (dec.) 存储条件: Ambient |
MDL编号:
MFCD00002637 CAS号: 583-93-7 |
产品技术规格
Appearance | White to beige to tan Powder and/or chunks |
Infrared spectrum | Conforms |
Titration with HClO4 | 94.0 to 106.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...