Water purification systems for healthcare applications, BIOPURE 300/600
供应商: VEOLIA WATER TECHNOLOGIES
MB300XXM1-230 MB600XXM1-230
ELGAMB300XXM1-230EA 0 CNY
ELGAMB300XXM1-230 ELGAMB600XXM1-230
Water purification systems for healthcare applications, BIOPURE 300/600
超纯水系统 纯水和超纯水系统
The BIOPURE system features a unique, award winning design incorporating a very high output water purification system with a large 350 liter storage reservoir. The system is designed to feed large or multiple automated endoscope reprocessors and washer disinfector installations with compliant purified water.
  • Compact footprint to save space and facilitate easy installation
  • Guaranteed bacterial specification, provided by onboard technology combined with easy system disinfection
  • Reliable water quality – unique sanitisation design for purification of RO
More Product Information
现在订购
规格
型号 Biopure 300 Biopure 600
Environmentally Preferable
给水 Potable supply (tap water) Potable supply (tap water)
流速 30 l/min 30 l/min
细菌 <0,1 CFU/ml <0,1 CFU/ml
内毒素 <0,25 EU/ml <0,25 EU/ml
宽度×深度×高度 730×890×1820 mm 756×1100×1820 mm
重量 180 kg 210 kg