Buffers, running buffers for Biacore Systems
供应商: Cytiva
BR-1006-71 BR-1006-70 BR-1006-69 BR-1008-27 BR-1008-28
GEHEBR-1006-71EA 0 CNY
GEHEBR-1006-71 GEHEBR-1006-70 GEHEBR-1006-69 GEHEBR-1008-27 GEHEBR-1008-28
Buffers, running buffers for Biacore Systems
缓冲(液) 电泳缓冲液 Running Buffers
Buffers developed and verified to work in Biacore™ systems.
The recommended running buffer for an assay depends on the type of molecules used in the interaction, which assay will be run, and the type of sensor chip used. Supplied in ready to use or concentrated form.