Sand, quartz, 经纯化,清洗并点燃, J.T.Baker®

供应商: Avantor

BAKR3382-01EA 906 CNY
BAKR3382-01 BAKR3382-05
Sand, quartz, 经纯化,清洗并点燃, J.T.Baker®
Sand, quartz

Washed and ignited. For boats.

分子式: SiO₂
沸点: 2230 °C (1013 hPa)
溶点: 1427 °C
密度: 2,622…2.645 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD02100519
CAS号: 14808-60-7
EINECS: 238-878-4

现在订购

产品技术规格

For Laboratory,Research,or Manufacturing Use = 0.16 %
Substances Soluble in HCl ≤0.16 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR