Disinfectant tablets
供应商: VEOLIA WATER TECHNOLOGIES
CT3 CT3-50
ELGACT3EA 0 CNY
ELGACT3 ELGACT3-50
Disinfectant tablets
清洗剂 消毒剂
These disinfectant tablets sanitises tubing or pipe work to remove bacteria, reduce bio film and protect the system against bacterial growth.