Pipette Tips, Traditional
供应商: RAININ
RC-1000 30389279 30389186 17001122 30389281 30389283 30389287 30389289 17005939 17001975 17001126 17001117 30389163 30389164 30389168 30389181 30389183 30389185 30389189 30389191 30389193 30389199
RAINRC-1000EA 263.25 CNY
RAINRC-1000 RAIN30389279 RAIN30389186 RAIN17001122 RAIN30389281 RAIN30389283 RAIN30389287 RAIN30389289 RAIN17005939 RAIN17001975 RAIN17001126 RAIN17001117 RAIN30389163 RAIN30389164 RAIN30389168 RAIN30389181 RAIN30389183 RAIN30389185 RAIN30389189 RAIN30389191 RAIN30389193 RAIN30389199
Pipette Tips, Traditional
吸头
These thin-wall tips are designed to fit RAININ and Pipetman pipettes with conical shafts, Eppendorf and Labsystems pipettes. Some tips feature a hydrophobic filter which forms a barrier against aerosols and liquids eliminating potential sample and pipette cross contamination.
  • Filtered and pre-sterilised bioclean tips are gamma-irradiated
  • ShaftGard tips are longer than standard tips for easy access into microcentrifuge and PCR tubes
  • Wide orifice tips minimise shear forces on delicate samples and flow resistance for viscous samples
The tips areMore Product Information