安全开箱刀, Slice™
供应商: THE SAFETY KNIFE
SLICEBOXCUTTER SLICEAUTCUTTER
KNIFSLICEBOXCUTTEREA 0 CNY
KNIFSLICEBOXCUTTER KNIFSLICEAUTCUTTER
安全开箱刀, Slice™
刀和刀具
These cutters feature corrosion-resistant double-sided blades made ceramic, of one of the most durable materials on earth. Available with an auto or manual retractable blade.
  • Stays sharp up to 10× longer than steel
  • Easy access blade storage
  • Replaceable blades with safer rounded tip
  • Storage hook handle also protects hands
  • Available auto-retractable for added safety
  • Oil-free finish is perfect for cleanrooms
Blade: Zirconium oxide ceramic
Body: ABS plastic