Carbon steel or stainless steel blade, ABS plastic body. The Swan 300 is a disposable safety knife with push and pull action. It comes complete with a tape cutter and opposing slopes at the blade exit for a more efficient cut.
- No removable parts
- Pointed nose for puncturing into material: Ideal for bag and sack cutting
- Opposing slopes at the blade exit reduces friction for a more efficient cut
- With handle grip and lanyard point
- With stainless steel tape cutter
- Available as metal detectable version with stainless steel blade
Ideal for cutting stretch / bubble wrap, strapping/banding, card/tape, multi layer bags/sacks, string and netting.