VWR®, Cleanroom Spiral Notebooks
供应商: VWR BRAND PRIVATE LABEL USA
89041-192 89041-194 89041-188 89041-194_U 89041-192_U 89041-188_U
VWRU89041-192EA 0 CNY
VWRU89041-192 VWRU89041-194 VWRU89041-188 VWRU89041-194_U VWRU89041-192_U VWRU89041-188_U
VWR®, Cleanroom Spiral Notebooks
笔记本
These spiral-bound notebooks feature latex paper in college-rule or 6,4 mm grid styles. Notebooks have 100 to 200 pages each.
  • Fully recyclable
  • Covers are of semi-transparent, high density polyethylene