VWR®, Aluminium Foil Seals for Cold Storage
供应商: VWR BRAND PRIVATE LABEL USA
89049-034 60941-112
VWRU89049-034EA 0 CNY
VWRU89049-034 VWRU60941-112
VWR®, Aluminium Foil Seals for Cold Storage
密封件 微孔板封板膜和铝箔
Aluminium foils with biologically inert adhesive. These foils are easily pierced by manual or automated pipette tips, for sample recovery without generation of aerosols or cross-contamination.
  • Recommended for temperatures from −80 to +130 °C
  • Not recommended for PCR
Each foil L×D: 133×82 mm