VWR® Talon® , Cylinder wall brackets
供应商: VWR BRAND PRIVATE LABEL USA
60142-004 60141-124
VWRU60142-004EA 0 CNY
VWRU60142-004 VWRU60141-124
VWR® Talon® , Cylinder wall brackets
墙钩
Cast aluminium cylinder wall mount brackets with strap allow cylinders to fit firmly along the support edge.
Features a 25 mm wide, 1372 mm long nylon strap with nickel plated, non slip spring catch and buckle for fast, easy adjustment.