VWR® Pipette Pump™ , Pipetting devices
供应商: VWR BRAND PRIVATE LABEL USA
53502-222 53502-233 53502-244
VWRU53502-222EA 1154.6 CNY
VWRU53502-222 VWRU53502-233 VWRU53502-244
VWR® Pipette Pump™ , Pipetting devices
移液管和移液器 移液管移液器
These pipetting devices accept both disposable and standard pipettes in the flexible tapered chuck, per ASTM Standard E714-80. They provide safe, accurate and trouble-free pipetting with simple, one-handed operation.
  • Pipettes fit smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger may be pressed for quick emptying
  • Resistant to acids and alkalis
  • Disassemble all parts for cleaning and before autoclaving at 121 °C (250 °F) for 20 minutes; chuck should not be autoclaved
  • Colour-coded for quick identification
订购须知 For replacement pipettor chucks, please contact your Avantor representative.