VWR® Talon®, Hosecock clamps, regular
供应商: VWR BRAND PRIVATE LABEL USA
21716-102 89202-614
VWRU21716-102EA 328 CNY
VWRU21716-102 VWRU89202-614
VWR® Talon®, Hosecock clamps, regular
夹具 油管夹
Hosecock clamps deliver precise flow control for tubing up to 17 mm diameter.
  • Adjustment screw has an oversized head for easy one-hand operation and accurate regulation of fluid flow
  • Tubing retainer screw on side of clamp keeps tubing from disengaging under varying fluid pressures, and can be raised to allow easy positioning of tubing
  • Convex bearing surface reduces the amount of pressure required for complete closure; eliminates sharp edges that could cut into tubing