VWR® Talon®, S-connectors
供应商: VWR BRAND PRIVATE LABEL USA
60097-056 12621-246
VWRU60097-056EA 250 CNY
VWRU60097-056 VWRU12621-246
VWR® Talon®, S-connectors
夹具 锤头
S-connectors made from electro-polished stainless steel or nickel plated zinc.
  • Clamp is ideal for constructing lab frames or other supports requiring the connection of two perpendicular rods
  • Clamp connects two 13 mm rods at a 90° angle and features separate adjustment screws for each rod location