VWR®, Water Sampling Bottles, Amber, Boston Type, Round
供应商: VWR BRAND PRIVATE LABEL USA
89093-892 89093-894 89093-908
VWRU89093-892EA 1588 CNY
VWRU89093-892 VWRU89093-894 VWRU89093-908
VWR®, Water Sampling Bottles, Amber, Boston Type, Round
水取样瓶
Type III (soda-lime) glass, amber.
  • Open-top cap with ultrasonically bonded fluoropolymer resin/silicone septa make them ideal for use in volatile organic analysis
  • Amber glass is recommended for light sensitive samples
Quality-Assured® (QA) packers follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. Not Processed (NPC) packers do not receive the specialty cleaning process.
Delivery information: QA packers include certificate of analysis, bar codes with lot number and individual container number for complete traceability.