VWR® Talon® , Retort rods
供应商: VWR BRAND PRIVATE LABEL USA
60079-381 60079-440 60079-428 60079-360
VWRU60079-381EA 201 CNY
VWRU60079-381 VWRU60079-440 VWRU60079-428 VWRU60079-360
VWR® Talon® , Retort rods
搅拌棒
Aluminium. For use with VWR® Talon® lab-frames.
  • Smooth fit with lab frames
  • Center-less ground for precise fit
  • Corrosion-resistant with chamfered ends for easy location