VWR® Talon®, Retort rings, open
供应商: VWR BRAND PRIVATE LABEL USA
28641-012 28641-010
VWRU28641-012EA 0 CNY
VWRU28641-012 VWRU28641-010
VWR® Talon®, Retort rings, open
曲颈瓶环
These lightweight rings mount funnels, boiling flasks, and other irregular-shaped objects to lab frames.
  • Open ring section allows items to pass in and out of support area easily, reducing risk of breakage
  • Mount to rods and frames with connectors such as VWR® Talon® regular or heavy-duty holders
Extension arm length: 57 mm