VWR®, 该性能均质袋
供应商: VWR
VWR®
VWRI129-1105EA 0 CNY
VWRI129-1105 VWRI129-1106 VWRI129-1103 VWRI129-1104 VWRI129-1102
VWR®, 该性能均质袋
混合器袋
Complete range of bags, sterile and alimentary certified, for safe and quick preparation of solid samples before microbiological and chemical analysis. Bags are resistant for three minutes blending (frequency 4 t/second). They fit all conventional 400 ml laboratory blenders, and are adapted to all samples.
  • Can resist to high temperature from –40 to +80 °C
  • Can be frozen
  • Gamma irradiated: No risk of cross-contamination (the red sticker on every box serves as a qualitative indicator of gamma ray processing)
  • Colour-change label when sterile: Red colour, for easy sterilisation identification
  • Approved for food contact
Plain blender bag: Sterile, flexible and transparent bag compatible with any laboratory blender. High quality of raw material: Pure natural polyolefin (polyethylene).
  • 可耐受– 40 to + 80 °C温度
  • 可冷冻
  • 可经γ辐照:无交叉感染风险(每个盒子上的红色贴纸可作为γ射线处理的定性指标)
  • 灭菌时贴纸的颜色便发生变化:红色便于灭菌识别
  • 经批准可接触食品
Bag with lateral filter: Non woven filter with porosity <250 µm in a reinforced multi-coated complex which is ideal for homogenising fibrous samples.
Bag with full page filter: Calibrated micro perforated filter with 280 µm porosity which is ideal for homogenising pasty samples.
符合ISO 7218 / 6887- 1 / FDA - BAM标准.
根据GMP质量保证及现行法规的要求进行灭菌,同时,我们特此证明产品已经过γ射线处理.