VWR®, Inoculating Loops and Needles, Sterile
供应商: VWR BRAND PRIVATE LABEL USA
89126-870 89126-868
VWRU89126-870EA 1311 CNY
VWRU89126-870 VWRU89126-868
VWR®, Inoculating Loops and Needles, Sterile
涂布棒 接种环和针
PS, sterile. Loops and needles are smooth and flexible to facilitate uniform streaking without damaging the gel surface. Needles are straight and suitable for removal of specimens of single colonies.
  • Colour coded for easy size identification
  • Packaged sterile in safe, tamperproof, zip-seal resealable bags
  • Disposable loops and needles do not need flaming, no risk of infection due to improper sterilisation
  • Loops and needles can be used under hoods without danger
Packaging: Packed sterile in safe, tamperproof, zip-seal resealable bags.