VWR®, Hybridisation Ovens
供应商: VWR BRAND PRIVATE LABEL USA
97005-254 97019-064
VWRU97005-254EA 107675 CNY
VWRU97005-254 VWRU97019-064
VWR®, Hybridisation Ovens
干燥箱 杂交炉
These hybridisation ovens are high capacity units, that can accomodate many different sized vessels. Designed to provide rapid heat-up, excellent temperature stability and efficient mixing for all types of hybridisation and incubation applications.
  • Compact design
  • Forced air convention
  • Programmable digital timer, up to 99 hours 59 minutes