VWR®, Testing Sieves, All Brass
供应商: VWR BRAND PRIVATE LABEL USA
57334-106 57334-108 57334-124 57334-104 57334-112 57334-126 57334-132
VWRU57334-106EA 2466 CNY
VWRU57334-106 VWRU57334-108 VWRU57334-124 VWRU57334-104 VWRU57334-112 VWRU57334-126 VWRU57334-132
VWR®, Testing Sieves, All Brass
筛子 试验筛
Brass frame and woven wire cloth offers higher durability than all brass construction. Sieves feature sturdy construction with fillet solder bead between the frame and wire cloth to prevent particles from being trapped. Diameter: 20,3 cm (8"). Choose full height with 5,08 cm (2") depth to wire.
Inspected with an optical measurement system traceable to the National Institute of Standards and Technology. Each sieve bears a traceable serial number and comes with certificate of compliance to applicable ASTM E-11 and ISO specifications.