VWR®, Mortars without pestles, porcelain
供应商: VWR BRAND PRIVATE LABEL USA
89038-154 89038-148 89038-150 89038-152
VWRU89038-154EA 1476 CNY
VWRU89038-154 VWRU89038-148 VWRU89038-150 VWRU89038-152
VWR®, Mortars without pestles, porcelain
研钵和杵 研钵
Porcelain, glazed exterior, with spout.
订购须知 Pestle must be ordered separately.