VWR® Spinbar®, Magnetic stirring bars, micro
供应商: VWR BRAND PRIVATE LABEL USA
58948-375 58948-397 58948-353 58948-976 58948-377 58948-400 76001-872
VWRU58948-375EA 61 CNY
VWRU58948-375 VWRU58948-397 VWRU58948-353 VWRU58948-976 VWRU58948-377 VWRU58948-400 VWRU76001-872
VWR® Spinbar®, Magnetic stirring bars, micro
搅拌器 磁力搅拌子
Mainly designed for stirring small volumes in vessels such as vials, tubes and gradient makers.
  • Available in a variety of colours and sizes
  • Used in environmental testing and life science applications