VWR® Tubing, Rubber, Vacuum
供应商: VWR BRAND PRIVATE LABEL USA
62995-335 62993-690 62993-781 62993-726
VWRU62995-335EA 670 CNY
VWRU62995-335 VWRU62993-690 VWRU62993-781 VWRU62993-726
VWR® Tubing, Rubber, Vacuum
胶管 Transfer Tubing
Natural rubber, black/red.
  • Flexible, very good tensile strength
  • Seamless and acid-proof
  • It also offers high resilience and tear resistance
Heavy wall, suitable for vacuum applications in the laboratory.
Hardness (Shore durometer A): 50
Tensile strength: 172 to 241 bar (black) 138 to 179 bar (red)