VWR®, Crucibles, Porcelain
供应商: VWR BRAND PRIVATE LABEL USA
89038-000 89037-998 89037-994 89037-988 89038-034 89038-002
VWRU89038-000EA 701 CNY
VWRU89038-000 VWRU89037-998 VWRU89037-994 VWRU89037-988 VWRU89038-034 VWRU89038-002
VWR®, Crucibles, Porcelain
坩埚
Porcelain, glased inside and out, except base and rim.
  • Withstand maximum temperature of 1150 °C
  • Not recommended for minerals or metals
Lids are available separately.