VWR®, Tubing, PVC, Clear
供应商: VWR BRAND PRIVATE LABEL USA
89068-526 89068-534
VWRU89068-526EA 0 CNY
VWRU89068-526 VWRU89068-534
VWR®, Tubing, PVC, Clear
胶管 Transfer Tubing
PVC, clear, flexible, non toxic.
  • Lightweight
  • Resistant to chemicals and corrosion
  • Imparts no taste or odour to the critical stream
Ideal for use in general industrial, food, pharmaceutical, medical, chemical, and mechanical applications, includes protective coverings and insulation sleeving. Its smooth interior surface provides excellent flow characteristics.
Shore hardness: 68° (A)
Certifications: Meets USP Class VI; NSF 51 (food contact application), conforms to FDA and 3-A standards.