VWR®, Tubing, Silicone
供应商: VWR BRAND PRIVATE LABEL USA
89068-436 89068-468 89068-442 89068-454 89068-490 89068-456 89068-484
VWRU89068-436EA 3610.01 CNY
VWRU89068-436 VWRU89068-468 VWRU89068-442 VWRU89068-454 VWRU89068-490 VWRU89068-456 VWRU89068-484
VWR®, Tubing, Silicone
胶管 Transfer Tubing
Platinum-cured silicone tubing is flexible, durable, low volatile grade, and translucent. This tubing is ideal for a variety of transfer applications such as pharmaceutical, laboratory, food and beverage, and bioprocess manufacturing. It features an ultra-smooth interior surface that provides uniform fluid flow and resistance to bacteria build-up.
  • Can be sterilised by autoclaving and gamma-irradiation
  • Imparts no taste or odour to critical stream
Certifications: Meets USP Class VI; USP 661 (Physicochemical Tests); ISO10993, EP 3.1.9, 3-A standards andMore Product Information