VWR®, Aluminum foil, premium, ultra-clean
供应商: VWR BRAND PRIVATE LABEL USA
89107-724 89107-732 89107-736 89107-726 89107-730
VWRU89107-724EA 109 CNY
VWRU89107-724 VWRU89107-732 VWRU89107-736 VWRU89107-726 VWRU89107-730
VWR®, Aluminum foil, premium, ultra-clean
铝箔
Premium, ultra-clean aluminium foil is strong and puncture-resistant.
  • Available in a variety of formats
  • Contains 200 sheets
  • Puncture-resistant
Foil is able to withstand extreme heat and cold, and has the lowest moisture-vapour transfer rate of all wrapping materials. Standard, heavy-duty, and extra heavy-duty formats are supplied as boxed rolls with a metal tear-off bar for ease in obtaining exact size requirements. The pop-up dispenser contains 200 individual sheets.
Delivery information: Supplied as boxed rolls with a metal tear-off bar for easeMore Product Information