VWR®, Hydrometers
供应商: VWR BRAND PRIVATE LABEL USA
34610-002 34640-207 34623-150 34620-160 34615-006 34625-666 34623-106 34629-011
VWRU34610-002EA 591 CNY
VWRU34610-002 VWRU34640-207 VWRU34623-150 VWRU34620-160 VWRU34615-006 VWRU34625-666 VWRU34623-106 VWRU34629-011
VWR®, Hydrometers
液体比重计
These hydrometers are designed for determining the specific gravity of liquids, which are heavier than water.
  • Individually serialised
  • Feature a lead free metal ballast
  • Temperature of standardisation: 15,6 °C
For use in food and beverage, chemical, pharmacology, transportation, refrigeration.
Certifications: NIST and DAkkS. A2LA accreditions.
Delivery information: Supplied with multi-language statement of accuracy.