Safety knife, Shark
供应商: THE SAFETY KNIFE
SHARKMHOOKBLUE SHARKYLW-HOOK SHARKBLK-HOOK
KNIFSHARKMHOOKBLUEEA 0 CNY
KNIFSHARKMHOOKBLUE KNIFSHARKYLW-HOOK KNIFSHARKBLK-HOOK
Safety knife, Shark
刀和刀具
Stainless steel blade, glass filled nylon plastic body. Moulded version of the big fish safety knife with spring loaded hook blade and opposing slopes at the back of the blade exit for an efficient cut.
  • Mouth opening: 9 mm (cuts up to 12 mm rope)
  • Opposing slopes at the mouth exit reduces friction for a more efficient cut
  • Retractable hook blade which can be set to auto or manual retract
  • Carbon steel or stainless steel hook blade
  • Reversible and replaceable blades
Ideal for cutting strapping/banding, bubble wrap, tapes, cables, ropes, netting and rubber.