Quantitative migroorganisms, BioBall® SingleShot 30
供应商: BIOMERIEUX
56045
BMER56045EA
0
CNY
BMER56045
Quantitative migroorganisms, BioBall® SingleShot 30
微生物学测试系统
微生物检验
BioBall® contains a precise number of microorganisms in a water soluble ball delivering unprecedented accuracy for Quantitative Microbiological Quality Control. BioBall SingleShot – mean of between 28 and 33 cfu with a standard deviation of 3 cfu or less (unless otherwise stated). Designed for a precise single inoculation. Ideal positive control for media QC, positive control for pathogen testing in the food industry and as a positive control for water and waste water testing.
- Calibrated microbial reference
- No preparation time
- Easy to use : BioBall® dissolves instantly