Serological pipettes, Shorty, KIMBLE®
供应商: KIMBLEKONTES
72115-25210 72115-50510
KIML72115-25210EA 14440 CNY
KIML72115-25210 KIML72115-50510
Serological pipettes, Shorty, KIMBLE®
移液管和移液器 血清移液管
Borosilicate glass, disposable, sterile. These pipettes are ideal for use in tissue culture work and other procedures performed under a fume hood or in other confined areas.
  • 'Negative' graduations to increase the versatility of the pipette
  • Calibrated To Deliver (TD)
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Packaging: Wrapped in colour-coded paper/plastic packaging.