微量离心管,0.5/1.5/2.0 ml,SuperClear
供应商: VWR BRAND PRIVATE LABEL USA
89004-302 89004-300 89004-308
VWRU89004-302EA 3087 CNY
VWRU89004-302 VWRU89004-300 VWRU89004-308
微量离心管,0.5/1.5/2.0 ml,SuperClear
管子 微型离心管
超净PP树脂微量离心管,带刻度。
  • Can withstand centrifugation upto 20000×g
  • Caps include silicone O-rings for secure storage
  • Sterile tubes are sterilised by gamma irrardiation
  • Caps have smooth surface for applying labels
  • Sustainable design based on virgin resins
Packaging: 非无菌管和瓶盖包装在可重复密封的袋中。
带盖无菌管每50支包装于无菌袋中,每大包包含10袋。