Gas regulators
供应商: AGILENT HEWLETT PACKARD
0101-1536 0101-1535 0101-1537
HEWL0101-1536EA 0 CNY
HEWL0101-1536 HEWL0101-1535 HEWL0101-1537
Gas regulators
气体调节器
These gas regulators ensure that ICP-MS applications and methods are carried out with the consistent and stable gas flows. Effective gas regulation is vital for ensuring that ICP-MS systems operate from a stable basis allowing industry-leading levels of sensitivity to be achieved in relevant systems.
A range of specific models have been developed for various important gases and manufactured according to high standards.
现在订购
规格
类型 Two stage
Environmentally Preferable
最大入口压 207 bar
出口压力 1,03 bar