Surgical masks, PFR95* N95
供应商: KIMBERLY CLARK
46727 62126
KIMB46727EA 0 CNY
KIMB46727 KIMB62126
Surgical masks, PFR95* N95
口罩 外科口罩
Particulate filter respirator and surgical masks has a flat-fold duckbill design with pouch style.
  • Two polyurethane headbands secure the mask to face for tight facial seal
  • Soft lining and two levels of fluid protection
  • Large breathing chamber for added comfort
  • Provides a minimum of 95% filtration efficiency of 0,3 μm particles
Certifications: NIOSH approved