Volumetric flasks, class A, glass, clear, TS penny-head glass stopper
供应商: HIRSCHMANN
282-G-100 282-G-2000
HIRS282-G-100EA 0 CNY
HIRS282-G-100 HIRS282-G-2000
Volumetric flasks, class A, glass, clear, TS penny-head glass stopper
烧瓶 容量瓶
DURAN®, borosilicate glass, clear, with TS penny-head glass stopper.
  • Fine white graduation line and marking spot
Certification flasks comes with certificate of identification, accuracy and also with serial number which are laser printed onto its body.
Certifications: ASTM E288