VWR® Traceable® Ultra, 三显示温度计
供应商: VWR Collection
VWRI620-2334EA 2185 CNY
VWRI620-2334 VWRI620-2335 VWRI620-2336 VWRI620-2337 VWRI620-2338 VWRI620-2339 VWRI620-2340
VWR® Traceable® Ultra, 三显示温度计
温度计 数字温度计
Digital thermometers are designed to accurately monitor the temperature of fridges, freezers, incubators, water baths or during transport, in one or two different areas with enclosed temperature-buffered sensors or bullet sensors.
  • Smart alarm:持续可视报警和声音报警,即使装置已回到非报警条件
  • 三重显示能够同时显示最大/最小值和当前温度
  • 最大/最小值指示
Probes: 1 or 2 bullet sensors/1 or 2 bottle probes with non toxic glycol, in 2 versions; 5 ml bottle or micro bottle to simulate vaccines.
Certifications: 校准符合ISO/IEC 17025、ANSI/NCSL Z540-1和9001标准,经A2LA认证,NIST标准可追溯性。
Delivery information: 交付信息: 随附Traceable®校准证书、翻开式支架、墙壁安装件、VELCRO®、磁条和低电指示器。