Conductivity meter, handheld, sensION+ EC5
供应商: HACH
LPV3560970002 LPV3500970002
HACHLPV3560970002EA 0 CNY
HACHLPV3560970002 HACHLPV3500970002
Conductivity meter, handheld, sensION+ EC5
电化学仪器 电导率仪
This all-in-one system with guided menu navigation makes general water quality meter testing fast and simple. The meter is designed to be used in a wide variety of applications, primarily in the field.
  • Guided navigation menu allows you to follow simple prompts to set up your measurements thus reducing the time necessary to start testing, and improving the accuracy of measurements
  • Instrument is ergonomically shaped and can easily be operated single-handed
  • Designed for field use, operates on three AA batteries
  • IP67 - protected
  • Visual measurement time indicator
Delivery information: Meter only (without electrode) is supplied with user documentation but without electrodes or accessories.
The field kit includes: sensION+ EC5 meter, sensION+ 5060 platinum conductivity cell with temperature sensor, sensION+ portable field carrying case, 125 ml of 147 µS/cm, 1413 µS/cm and 12,88 mS/cm calibration standards, colour-coded screwable calibration tubes, sample and measurement containers, and user documentation.